ContactUsLinkComponent
Mouse Thrombin-Antithrombin Complex ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Thrombin-Antithrombin Complex in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA78845-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Thrombin-Antithrombin Complex ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Thrombin-Antithrombin Complex in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA78845-96
Supplier:  ANTIBODIES.COM