Human Retinoid X Receptor alpha/RXRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Retinoid X Receptor alpha/RXRA in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA7781-48
Supplier: Antibodies.com