ContactUsLinkComponent
Mouse LTB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse LTB in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6552-96
New Product
Mouse LTB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse LTB in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6552-96