ContactUsLinkComponent
Human Huntingtin Associated Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of human Huntingtin Associated Protein 1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7444-96
Supplier:  ANTIBODIES.COM
New Product
Human Huntingtin Associated Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of human Huntingtin Associated Protein 1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7444-96
Supplier:  ANTIBODIES.COM