ContactUsLinkComponent
Human GITR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human GITR in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7065-48
New Product
Human GITR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human GITR in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7065-48