ContactUsLinkComponent
Human Gastrin Releasing Peptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of human Gastrin Releasing Peptide in serum, plasma, tissue homogenates, cell lysates, cell cu 1 * 48 Tests
Catalog # ANTIA6989-48
Supplier:  Antibodies.com
New Product
Human Gastrin Releasing Peptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of human Gastrin Releasing Peptide in serum, plasma, tissue homogenates, cell lysates, cell cu 1 * 48 Tests
Catalog # ANTIA6989-48
Supplier:  Antibodies.com