Human FLCN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human FLCN in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6462-48
Supplier: ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.