ContactUsLinkComponent
Mouse Hormone Sensitive Lipase/HSL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of mouse Hormone Sensitive Lipase/HSL in serum, plasma, tissue homogenates or other biological 1 * 48 Tests
Catalog # ANTIA82429-48
Supplier:  Antibodies.com
New Product
Mouse Hormone Sensitive Lipase/HSL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of mouse Hormone Sensitive Lipase/HSL in serum, plasma, tissue homogenates or other biological 1 * 48 Tests
Catalog # ANTIA82429-48
Supplier:  Antibodies.com