ContactUsLinkComponent
Human IDUA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human IDUA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6577-96
Supplier:  ANTIBODIES.COM
New Product
Human IDUA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human IDUA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6577-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.