ContactUsLinkComponent
Human Calpain 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Calpain 3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6651-96
Supplier:  ANTIBODIES.COM
New Product
Human Calpain 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Calpain 3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6651-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.