ContactUsLinkComponent
Rat Kidney Androgen-Regulated Protein/KAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of rat Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenates, and 1 * 96 Tests
  ANTIA80323-96
New Product
Rat Kidney Androgen-Regulated Protein/KAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of rat Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenates, and 1 * 96 Tests
  ANTIA80323-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.