ContactUsLinkComponent
Human PAM/Peptidyl-Glycine alpha-Amidating Monooxygenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PAM/Peptidyl-Glycine alpha-Amidating Monooxygenase in serum, pl 1 * 96 Tests
Catalog # ANTIA6459-96
Supplier:  ANTIBODIES.COM
New Product
Human PAM/Peptidyl-Glycine alpha-Amidating Monooxygenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PAM/Peptidyl-Glycine alpha-Amidating Monooxygenase in serum, pl 1 * 96 Tests
Catalog # ANTIA6459-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.