Human Ryanodine Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Ryanodine Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
ANTIA7411-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.