ContactUsLinkComponent
Human Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of human Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA8118-96
Supplier:  ANTIBODIES.COM
New Product
Human Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of human Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA8118-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.