Human Free Prostate Specific Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Free Prostate Specific Antigen in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
ANTIA76569-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.