ContactUsLinkComponent
Human AMIGO1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AMIGO1 in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA7739-96
New Product
Human AMIGO1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AMIGO1 in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA7739-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.