ContactUsLinkComponent
Rat SPON2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SPON2 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7842-96
Supplier:  ANTIBODIES.COM
New Product
Rat SPON2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SPON2 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7842-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.