ContactUsLinkComponent
Human Urocortin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Urocortin 3 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA6657-96
New Product
Human Urocortin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Urocortin 3 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA6657-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.