Rat Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA7898-48
Supplier: Antibodies.com