ContactUsLinkComponent
Human Guanylate Cyclase Activator 2B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Guanylate Cyclase Activator 2B in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA74812-96
Supplier:  ANTIBODIES.COM
New Product
Human Guanylate Cyclase Activator 2B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Guanylate Cyclase Activator 2B in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA74812-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.