Human SPARCL1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human SPARCL1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6513-96
Supplier: ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.