ContactUsLinkComponent
Human DHEA Sulfate ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative dete rmination of human DHEA Sulfate in serum, plasma, cell culture supernatant, cell or tissue lysate, other liquid sampl 1 * 96 Tests
Catalog # ANTIA74764-96
Supplier:  ANTIBODIES.COM
New Product
Human DHEA Sulfate ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative dete rmination of human DHEA Sulfate in serum, plasma, cell culture supernatant, cell or tissue lysate, other liquid sampl 1 * 96 Tests
Catalog # ANTIA74764-96
Supplier:  ANTIBODIES.COM