ContactUsLinkComponent
Human Melatonin Receptor 1B/MTNR1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Melatonin Receptor 1B/MTNR1B in tissue homogenates, cell lysates or other biological 1 * 48 Tests
Catalog # ANTIA7185-48
Supplier:  Antibodies.com
New Product
Human Melatonin Receptor 1B/MTNR1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Melatonin Receptor 1B/MTNR1B in tissue homogenates, cell lysates or other biological 1 * 48 Tests
Catalog # ANTIA7185-48
Supplier:  Antibodies.com