ContactUsLinkComponent
Human Proteasome 20S alpha 5/PSMA5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Proteasome 20S alpha 5/PSMA5 in tissue homogenates, cell lysates or other biological 1 * 48 Tests
Catalog # ANTIA7612-48
Supplier:  Antibodies.com
New Product
Human Proteasome 20S alpha 5/PSMA5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Proteasome 20S alpha 5/PSMA5 in tissue homogenates, cell lysates or other biological 1 * 48 Tests
Catalog # ANTIA7612-48
Supplier:  Antibodies.com