ContactUsLinkComponent
Rat Retinoic Acid Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of rat Retinoic Acid Receptor alpha in tissue homogenates or other biological fluids. 1 * 48 Tests
Rat Retinoic Acid Receptor alpha ELISA Kit
Catalog # ANTIA6265-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Rat Retinoic Acid Receptor alpha ELISA Kit
Catalog # ANTIA6265-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Some Products May Appear Restricted