ContactUsLinkComponent
Mouse Prion Protein PrP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of mouse Prion Protein PrP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Mouse Prion Protein PrP ELISA Kit
Catalog # ANTIA6205-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Mouse Prion Protein PrP ELISA Kit
Catalog # ANTIA6205-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Some Products May Appear Restricted