SYRINGE TEST HOLDER FOR TESTING PREFILLED SYRINGES 267635 | 90279000 | AT For syringes with outer diameter 6 to 22mm Test according to ISO 11040 and ISO 7886 1 * 1 items
Catalog # PAAR267635
Supplier: Anton Paar
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.