ContactUsLinkComponent
4-Hydroxymidazolam solution 1 mg/mL (90:10 Acetonitrile:DMSO), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-172-1ML
Supplier:  Merck
New Product
4-Hydroxymidazolam solution 1 mg/mL (90:10 Acetonitrile:DMSO), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-172-1ML
Supplier:  Merck