ContactUsLinkComponent
5-Hydroxyindole-3-acetic acid (5-HIAA) solution 1 mg/mL ( Degassed Methanol w/ 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant® 1 * 1 mL
Catalog # H-176-1ML
Supplier:  Merck
New Product
5-Hydroxyindole-3-acetic acid (5-HIAA) solution 1 mg/mL ( Degassed Methanol w/ 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant® 1 * 1 mL
Catalog # H-176-1ML
Supplier:  Merck