ContactUsLinkComponent
With Autofill Static Holding Time (days) - 173 Normal Evaporation Rate (liters/day) - 0.98 Liquid Nitrogen Capacity - 175 litres Neck Diameter (mm) - 216 Overall Height / Diameter (mm) - 963 x 673 1 * 1 items
Catalog # TAYW10-152-01-0055
Supplier:  TAYLOR WHARTON
New Product
With Autofill Static Holding Time (days) - 173 Normal Evaporation Rate (liters/day) - 0.98 Liquid Nitrogen Capacity - 175 litres Neck Diameter (mm) - 216 Overall Height / Diameter (mm) - 963 x 673 1 * 1 items
Catalog # TAYW10-152-01-0055
Supplier:  TAYLOR WHARTON