4,6-Dichloronicotinamide
Bottle
Catalog # APOSOR303826-250MG
New Product
Supplier: Apollo Scientific
4,6-Dichloronicotinamide
Catalog # APOSOR303826-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 70593-57-6
Jump to:
Product Details & Documents
- CAS Number:70593-57-6
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD01646073
- Molecular Weight:191.0200 g/mol
- Melting Pt:153…155 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



