Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Hexatriacontane 98%

Hexatriacontane 98%

Bottle
Catalog # APOSOR95695-100G
Supplier:   Apollo Scientific
Hexatriacontane 98%
Catalog # APOSOR95695-100G
Supplier:  Apollo Scientific
CAS Number:  630-06-8
Hexatriacontane 98%

Bottle

CAS Number:  630-06-8
Price
£534.00
Log in for availability
1 * 100 g
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Hexatriacontane
  • CAS Number:630-06-8
  • Formula:C₃₆H₇₄
  • MDL Number:MFCD00009413
  • Molecular Weight:506,97 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.