ContactUsLinkComponent
Exceptional results and smart quality assurance. 20.1 kg / 4.2 kg readability: 0.1 g / 0.01 g integrated methods results notepad SmartPan Pro weighing pan user passwords 4 x USB 1 x LAN error-free data 1 * 1 items
Catalog # METB30317528
Supplier:  Mettler - Toledo
New Product
Exceptional results and smart quality assurance. 20.1 kg / 4.2 kg readability: 0.1 g / 0.01 g integrated methods results notepad SmartPan Pro weighing pan user passwords 4 x USB 1 x LAN error-free data 1 * 1 items
Catalog # METB30317528
Supplier:  Mettler - Toledo