LANATOSID C, ROTICHROM® CHR, 10 MG, GLAS 1 * 10 mg
Catalog # ROTH1N3P.1
Supplier: Roth Carl
New Product
LANATOSID C, ROTICHROM® CHR, 10 MG, GLAS 1 * 10 mg
Catalog # ROTH1N3P.1
Supplier: Roth Carl
New Product
Frequently Bought Together
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Supplier: VWR Chemicals
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane for analysis
Supplier: Thermo Fisher Scientific