ContactUsLinkComponent
,,5 components: P 1000mg/l K 1000mg/l Mg 1000mg/l S 1000mg/l Na 500mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM096010L5
New Product
,,5 components: P 1000mg/l K 1000mg/l Mg 1000mg/l S 1000mg/l Na 500mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM096010L5