ContactUsLinkComponent
Funnel Universal, made of PE-EX, screw cap made of yellow PPS female thread: 8x UNF 1/4 28G (inclined), 4 x NPT 1/8 (straight) head diameter 56 mm total length 141 mm. Accessories: Fittings made of PFA (8x green for tubing O.D. 1,6 mm, 8x violet 1 * 1 items
Catalog # BOHLR803-04
Supplier:  Bohlender
New Product
Funnel Universal, made of PE-EX, screw cap made of yellow PPS female thread: 8x UNF 1/4 28G (inclined), 4 x NPT 1/8 (straight) head diameter 56 mm total length 141 mm. Accessories: Fittings made of PFA (8x green for tubing O.D. 1,6 mm, 8x violet 1 * 1 items
Catalog # BOHLR803-04
Supplier:  Bohlender