ContactUsLinkComponent
Calibration Standard for testing the wavelength accuracy, certified at:wavelengths: 241, 287, 354, 416, 444, 575, 641, 732, 740, 864nm slit width: 1nm 1 * 1 items
Catalog # HELL1X667045
Supplier:  HELLMA
New Product
Calibration Standard for testing the wavelength accuracy, certified at:wavelengths: 241, 287, 354, 416, 444, 575, 641, 732, 740, 864nm slit width: 1nm 1 * 1 items
Catalog # HELL1X667045
Supplier:  HELLMA