CELLCYTE 3 Inverted | Enhanced Phase Contrast | 4-Channel Fluorescence DAPI - EX:395/25 EM:450/30 | GFP - EX:483/18 EM:525/25 | RFP - EX:555/20 EM:595/30 | CY5 - EX:635/20 EM:695/60 | (Control Unit and Software (ECCC3CU3) ordered separately) 1 * 1 items
ECHLECCC3243
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.