Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Upper basket A 102 Height adjustable, for the optimum loading of inserts 1 * 1 items
Catalog # MIEL12645600
Supplier:
MIELE
New Product
Expand image
Upper basket A 102 Height adjustable, for the optimum loading of inserts 1 * 1 items
Catalog # MIEL12645600
Supplier:
MIELE
New Product
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% Ph. Eur., USP
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Lower basket A 151 for holding standard DIN mesh trays as well as various inserts. 1 * 1 items
Supplier: MIELE
Jerricans, Narrow Neck, HDPE
Supplier: RIXIUS