ContactUsLinkComponent
NEXOPART Air jet sieve machine eControl Basic Technical specifications: 90-264 V, 50/60 Hz 1500-5000 Pa Underpressure Dimensions L x W x H (mm): 535 x 370 x 386 Scope of delivery: softwareversion HMI 3.0 Basic, sieve adapter, slottet nozzle, acrylic 1 * 1 items
Catalog # HAVE211160814
Supplier:  VWR Collection
New Product
undefined
NEXOPART Air jet sieve machine eControl Basic Technical specifications: 90-264 V, 50/60 Hz 1500-5000 Pa Underpressure Dimensions L x W x H (mm): 535 x 370 x 386 Scope of delivery: softwareversion HMI 3.0 Basic, sieve adapter, slottet nozzle, acrylic 1 * 1 items
Catalog # HAVE211160814
Supplier:  VWR Collection