NEEDLE SPINAL QUINCKE 25GX2 0,5X50MM 1 * 25 items
Catalog # BDAM405245
Supplier: BD Medical
New Product
NEEDLE SPINAL QUINCKE 25GX2 0,5X50MM 1 * 25 items
Catalog # BDAM405245
Supplier: BD Medical
New Product
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Measuring scoops
Supplier: BURKLE
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
NEEDLE SPINAL YALE 25GX3 0,5X75MM 1 * 25 items
Supplier: BD Medical