ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1C1P Detection: S Back panel: Transparent Front and Side panel: Reverso + standard Worksurface: Stainless steel Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA666F01024D31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1C1P Detection: S Back panel: Transparent Front and Side panel: Reverso + standard Worksurface: Stainless steel Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA666F01024D31
Supplier:  ERLAB CAPTAIRE