ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1P Back panel: Standard Front and Side pa nel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA666E00014A31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1P Back panel: Standard Front and Side pa nel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA666E00014A31
Supplier:  ERLAB CAPTAIRE