ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1P2C Detection: F Back panel: Transparent Front and Side panel: Reverso + left waste port Power supply: GB Version: 1 1 * 1 items
  ERLAAA666D03026A31
New Product
Model technology: Filtration ductless fume hood Captair 714 Smart Filters configuration: 1P2C Detection: F Back panel: Transparent Front and Side panel: Reverso + left waste port Power supply: GB Version: 1 1 * 1 items
  ERLAAA666D03026A31