ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Detection: F Back panel: Transparent Front and Side panel: Reverso + right waste port Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C03025A31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Detection: F Back panel: Transparent Front and Side panel: Reverso + right waste port Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C03025A31
Supplier:  ERLAB CAPTAIRE