ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Detection: F Back panel: Standard Fr ont and Side panel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C03014A31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Detection: F Back panel: Standard Fr ont and Side panel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C03014A31
Supplier:  ERLAB CAPTAIRE