ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Back panel: Transparent Front and Si de panel: Sequential opening+ standard Worksurface: Trespa Toplab PLUS Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C00021C31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1P1C Back panel: Transparent Front and Si de panel: Sequential opening+ standard Worksurface: Trespa Toplab PLUS Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA646C00021C31
Supplier:  ERLAB CAPTAIRE