ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1C Detection: A Back panel: Transparent F ront and Side panel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
  ERLAAA646A02024A31
New Product
Model technology: Filtration ductless fume hood Captair 483 Smart Filters configuration: 1C Detection: A Back panel: Transparent F ront and Side panel: Reverso + standard Power supply: GB Version: 1 1 * 1 items
  ERLAAA646A02024A31