ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 2P1C Detection: S Back panel: Standard Fr ont and Side panel: Reverso + standard Worksurface: Trespa Toplab PLUS Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636J01014C31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 2P1C Detection: S Back panel: Standard Fr ont and Side panel: Reverso + standard Worksurface: Trespa Toplab PLUS Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636J01014C31
Supplier:  ERLAB CAPTAIRE