ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 1P1C1P Detection: A Back panel: Standard Front and Side panel: Sequential opening+ standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636I02011A31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 1P1C1P Detection: A Back panel: Standard Front and Side panel: Sequential opening+ standard Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636I02011A31
Supplier:  ERLAB CAPTAIRE